Learn More
High-Tech Conversions Virtu-Clean Class 100 Compatible Powder Free Nitrile Examination Gloves
Description
Virtu-Clean™ Gloves are ambidextrous gloves that are latex free, powder free, and a 12 in. cuff for critical environments. These gloves protect against the harmful effects of hazardous drugs.
- ISO Class 5 / Class 100 Compatible
- Non-sterile
- Powder-free; textured fingers
- USP <800> Compliant
- 100% Nitrile; Latex Free
- 12 in. long for added safety
Specifications
Specifications
| Chlorinated | No |
| Cleanroom Class | Class 100 (ISO 5) |
| Accelerator Free | Yes |
| AQL | 1.5 |
| Color | White |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 5 mil |
| Finger Thickness (Metric) | 0.13 mm |
| Hand | Ambidextrous |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.