Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio VirusGEN AAV Transfection Kit with RevIT AAV Enhancer

Catalog No. mir8007 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Volume (Metric):
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
MIR8007 1 L
MIR8008 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR8007 Supplier Mirus Bio Supplier No. MIR 8007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paired with TransIT VirusGEN, the RevIT AAV Enhancer delivers higher titers for recombinant adeno-associated virus production

  • TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • Kit includes: TransIT-VirusGEN Transfection Reagent and RevIT AAV Enhancer 
  • Key benefits: 
    • Performance: Efficient DNA delivery for production of high-titer viral vectors
    • Scalability: Efficient production from less than 1 mL to large scale culturesFlexibility: Compatible with different recombinant virus and cell culture systems
    • Animal Origin Free: Fully synthetic transfection reagent formulation

Specifications

Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Product Type AAV Transfection Kit with Enhancer
Sterility Sterile
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.