Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio VirusGEN™ AAV Transfection Kit

Catalog No. MIR6750 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6750 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR6750 Supplier Mirus Bio Supplier No. MIR 6750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VirusGEN™ AAV Transfection Kit is ideal for Recombinant Adeno-associated Virus and Lentivirus Production.

  • TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • For suspension 293 cells.
  • Kit includes: TransIT-VirusGEN Transfection Reagent (2 x 1.5 mL), VirusGEN AAV Complex formation Solution and Enhancer (100 mL)
  • Key benefits of TransIT-VirusGEN Transfection Reagent include:
  • Performance: Efficient DNA delivery for production of high-titer viral vectors
  • Scalability: Efficient production from < 1 ml to large scale cultures
  • Flexibility: Compatible with different recombinant virus and cell culture systems
  • Animal Origin Free: Fully synthetic transfection reagent formulation
  • TransIT-VirusGEN™ is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN AAV Transfection Kit and VirusGEN LV Transfection Kit.

Specifications

Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.