Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio VirusGEN™ LV Transfection Kit, 30mL

Catalog No. MIR6765 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6765 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6765 Supplier Mirus Bio Supplier No. MIR 6765
Add to Cart
Add to Cart
Request a Quote

Ideal for Lentivirus Production

  • TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • Kit includes: TransIT-VirusGEN Transfection Reagent , VirusGEN LV Complex formation Solution and VirusGEN LV Enhancer
  • TransIT-VirusGEN™ is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN AAV Transfection Kit and VirusGEN LV Transfection Kit.
  • Key benefits:
    • Performance: Efficient DNA delivery for production of high-titer viral vector
    • Scalability: Efficient production from less than 1 mL to large scale cultures
    • Flexibility: Compatible with different recombinant virus and cell culture systems
    • Animal Origin Free: Fully synthetic transfection reagent formulation

Specifications

Cell Type 293-derived Cells
Format Bottle
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Product Type LV Transfection Kit
Quantity 30 mL
Sterility Sterile
Volume (Metric) 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.