Learn More
Anton Paar ViscoQC 100 – R a single point medium viscosity rotational viscometer
Supplier: Anton Paar 105021
ViscoQC 100 is a digital rotational viscometer that measures the dynamic viscosity as a single-point value. Unique benefits include automatic spindle detection by Toolmaster™, automatic guard detection by TruGuard™, unique TruMode™, magnetic coupling for easy one-hand exchange of spindles and digital leveling to ensure the proper alignment of the instrument. It has no data memory for storing results. Continuous data output to a printer or via V-Collect software installed on a PC is possible.Speed range: 0.1 to 200 rpm (19 standard speeds plus 6 freely selectable speeds)Accuracy: ±1.0 % full scale rangeRepeatability: ±0.2 %R model is perfectly suited for medium-viscosity samples such as creams, food and paints. Viscosity range: 10* to 40M mPa·s (*with optional double-gap system DG26, with RH1 spindle 100 mPa·s).Includes: Fully assembled instrument, 6 spindles equipped with Toolmaster™, Magnetic coupling, SOP, V-Collect