Learn More
Koehler™ Instrument Viscosity Reference Standards

Description
For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic
• Standards are based upon the NIST value of 1.0034 cSt for water at 20°C (68°F)
• Traceable to National Standards; manufactured according to ASTM D2162 specifications
• Supplied with ISO/IEC 17025 Certification Report
• Fully compliant with ASTM and related test procedures
• Calibrated to a precision of ±0.2% for viscosity and kinematic viscosity
• Each standard is calibrated at a minimum of five temperatures
• Supplied in 500mL amber bottle, with full certification and MSDS
Specifications
Specifications
| Viscosity | 5.7 cSt at 40°C |
| Physical Form | Liquid |
| Technique | Calibration and Verification of Kinematic and Dynamic Viscosity Test |
| Certifications/Compliance | ISO/IEC 17025, ASTM D 2162, NIST |
| For Use With (Equipment) | Kinematic and Dynamic Viscosity Test Equipment |
| Type | Viscosity Standard |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.