Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Instrument Viscosity Reference Standards
SDP

Catalog No. 1361760
Encompass
Encompass_Preferred
Catalog No. 13-617-60 Supplier Koehler™ Instrument Supplier No. 355002060
Add to Cart
Add to Cart

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

• Standards are based upon the NIST value of 1.0034 cSt for water at 20°C (68°F)
• Traceable to National Standards; manufactured according to ASTM D2162 specifications
• Supplied with ISO/IEC 17025 Certification Report
• Fully compliant with ASTM and related test procedures
• Calibrated to a precision of ±0.2% for viscosity and kinematic viscosity
• Each standard is calibrated at a minimum of five temperatures
• Supplied in 500mL amber bottle, with full certification and MSDS

Specifications

Viscosity 60 cSt at 40°C
Physical Form Liquid
Technique Calibration and Verification of Kinematic and Dynamic Viscosity Test
Certifications/Compliance ISO/IEC 17025, ASTM D 2162, NIST
For Use With (Equipment) Kinematic and Dynamic Viscosity Test Equipment
Type Viscosity Standard
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.