Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ViscoTip™ - Syringe Tip
GREENER_CHOICE

Catalog No. 05413573 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-413-573 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-413-573 Supplier Eppendorf™ Supplier No. 0030089502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eppendorf ViscoTip™ syringe tips in 'Eppendorf Quality' grade excel in handling viscous liquids with unmatched precision. Ideal for biochemistry and molecular biology, they ensure minimal sample retention and ergonomic efficiency in laboratory tasks.

  • Specialized for liquids with a dynamic viscosity from 200 mPa*s to 14,000 mPa*s
  • For dispensing volumes from 100 μL to 10 mL in increments of 10 μL
  • Make life easier for your instrument: The new design of the tip means significantly lower operation forces are used, thus it speeds up work and reduces energy consumption
  • Don't waste time calculating: Automatic tip recognition and volume calculation in Repeater™ M4/E3/E3x
  • Free of leachables and slip agents: The ViscoTip is free of experiment-interfering leachables (e.g. slip agents, biocides, plasticisers)
  • Save time finding the right tip: The color coded double ring and the continuous volume scale distinguish the ViscoTip from Combitips™ advanced
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 mL
Volume (Metric) Dispensing 10 mL
Sterility Non-sterile
Clean Claims Eppendorf Quality (Non-sterile)
For Use With (Application) Highly viscous liquids
Packaging 100 Pk. (4 Bags x 25 Pk.)
Quantity 100 Pk. (4 Bags of 25)
Product Type Syringe Tip
Color Code Orange
For Use With (Equipment) MultiPipette
Graduated Yes
Product Line ViscoTip
Graduations 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.