Learn More
MilliporeSigma™ Supelco™ Visidry™ Drying Attachment
The Visidry Drying Attachment was designed for the Visiprep Vacuum Manifold (Cat. No. 57100-U also fits the Preppy manifold).
Supplier:  MilliporeSigma™ Supelco™ 57124
 View more versions of this product
Catalog No. 11-100-7063
Only null left
Add to Cart
Description
Description
The Visidry unit installs in minutes, dries up 12 or up to 24 SPE tubes at one time, and can be used with any inert gas supply. It is also useful for evaporating and concentrating recovered samples. Gas flow to each port can be independently adjusted.Note: The Visidry drying attachment cannot be used to dry 12, 20, or 60mL SPE tubes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Food and Beverages
Visiprep 24-Port Model
Visidry
SPE Accessory
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Visidry is a trademark of Sigma-Aldrich Co. LLC