Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Catalog No. 14381502
Change view
Click to view available options
Type:
12-user Software License
6-user Software License
VISIONlite 5 Software
VISIONlite 5 Software + Cable
VISIONlite ColorCalc Software
Wine Analysis Software
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-381-502 12-user Software License
14-381-500 VISIONlite 5 Software
14-380-015 6-user Software License
14-385-393 VISIONlite 5 Software + Cable
14-380-013 VISIONlite ColorCalc Software
14-387-623 Wine Analysis Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-381-502 Supplier Thermo Scientific™ Supplier No. 699130000
Add to Cart
Add to Cart
Request a Quote
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite 5 Software increases the capability of your instrument:
  • Add method and data storage for SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
  • Report results to 0.5nm wavelength resolution and 0.001A for SPECTRONIC 200 Spectrophotometer.
  • Perform automated kinetics measurements on SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
  • Scan on GENESYS 20 Visible Spectrophotometer.
  • Produce formatted reports in hard  copy or pdf format.
VISIONlite ColorCalc Software turns your GENESYS 10S UV-Vis Spectrophotometer or Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer  into a powerful color analyser:
  • The Basic package offers all the most common colorspaces, illuminants and observer angles.
  • The Advanced package offers specialized calculations for yellowness, pharmaceuticals and quality control in wine production.
TRUSTED_SUSTAINABILITY
Specifications
Type 12-user Software License
For Use With (Equipment) VISIONlite 5 Software
Includes Includes two copies of the installation disc and a license granting approval for twelve installed instances of the software