Learn More
Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader
Description
Decode a full rack of barcoded storage tubes in less than ten seconds with the Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader. This compact, user-friendly reader provides exceptional flexibility with data export formats, tube type readability, and tube layout options.
- Controlled via on-screen software interface or remotely via TCP/IP connection
- Scans a full tube rack in less than ten seconds
- Simply follow the instructions during software installation and connect the reader to a computer via USB port
- Integrates easily with LIMS, existing databases or tracking systems
- Small footprint and compact profile save valuable laboratory space
Specifications
Specifications
| Description | VisionMate SR 2-D Bar Code Reader |
| Decoding Capacity | Single Rack |
| Data Transfer Method | Software Interface or Remotely via TCP/IP Connection |
| Speed | Full tube rack in less than ten seconds |
| Type | Barcode Reader |