Learn More
Medchemexpress LLC HY-10440 200mg , Vismodegib GDC-0449 CAS:879085-55-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10440200MG
Encompass_Preferred
Medchemexpress, HY-10440 200mg Vismodegib GDC-0449 CAS:879085-55-9 Formula:C19H14Cl2N2O3S IC50: 3 nM (hedgehog), 3.0 μM (P-gp), 1.4 μM (ABCG2) Purity:98% Vismodegib is an orally active hedgehog pathway inhibitor with an IC 50 of 3 nM and also inhibits P-gp, ABCG2 with IC 50 values of 3.0 μM and 1.4 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2574
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.