Learn More
Enzo Life Sciences Vismodegib (free base) (5g). CAS: 879085-55-9
SDP
Supplier:  Enzo Life Sciences ENZCHM2425000
Encompass_Preferred
Vismodegib is a potent, novel and specific inhibitor of hedgehog with IC50 of 3 nM and also inhibitor of P-gp with IC50 of 3.0 μM. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO at 200 mg/mL. Long Term Storage: -20°C.
Catalog No. 50-201-3699
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.