Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Vismodegib Hedgehog inhibitor
SDP

Catalog No. NBP2627585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2627585 5 mg
NBP26275825 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2627585 Supplier Novus Biologicals™ Supplier No. NBP2627585MG
Add to Cart
Add to Cart

May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)

  • Purity:>98% (HPLC); NMR (conforms)
  • Molecular Weight: 421.3
  • PubChem ID:24776445

Specifications

CAS 879085-55-9
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Donor Source Synthetic
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.