Learn More
CTL Scientific Supply Corp VISOCOLOR ECO PH 4.0-9.0
SDP
Supplier:  CTL Scientific Supply Corp 931066
Encompass_Preferred
Environmental test kit for pH Gradation of scale: 4.0 - 5.0 - 6.0 - 6.5 - 7.0 - 7.5 - 8.5 - 8.5 - 9.0 mg/l 400 tests/kit
Catalog No. 50-300-381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.