Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Reagent Reservoirs
SDP

Catalog No. 21385124 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Capacity (Metric):
10 mL
100 mL
100 mL (with lid)
1800 mL
25 mL
25 mL (divided)
50 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Individually packed
Quantity:
10/bag x 10; 100/Cs.
100/Cs.
20/bag x 10; 200/Cs.
200 in. 20 bags of 10
50/Cs.
Each
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Packaging Quantity
21-385-124 1800 mL Sterile Individually Wrapped Each
21-381-27A 100 mL Sterile Bulk 10/bag x 10; 100/Cs.
21-381-27B 100 mL Sterile Individually Wrapped 50/Cs.
21-381-27E 25 mL (divided) Sterile Bulk 10/bag x 10; 100/Cs.
21-385-111 100 mL (with lid) Sterile Individually Wrapped 50/Cs.
21-381-27D 25 mL Sterile Individually Wrapped 50/Cs.
2138127G 50 mL Sterile Bulk 10/bag x 10; 100/Cs.
2138127H 50 mL Sterile Individually Wrapped 50/Cs.
21-385-101 100 mL Sterile Individually Wrapped 100/Cs.
21-385-100 100 mL Sterile Bulk 20/bag x 10; 200/Cs.
21-385-104 50 mL Sterile Bulk 20/bag x 10; 200/Cs.
21-385-102 25 mL Sterile Bulk 20/bag x 10; 200/Cs.
21-385-105 50 mL Sterile Individually Wrapped 100/Cs.
21-385-103 25 mL Sterile Individually Wrapped 100/Cs.
22-145-101 25 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
22-145-102 25 mL (divided) Non-sterile Bulk 10/bag x 10; 100/Cs.
22-145-103 50 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
22-145-100 100 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
21-381-27C 25 mL Sterile Bulk 10/bag x 10; 100/Cs.
21-381-27F 25 mL (divided) Sterile Individually Wrapped 50/Cs.
21-381-090 10 mL Sterile Bulk 100/Cs.
21-381-091 10 mL Sterile Individually packed 50/Cs.
21-381-092 10 mL Sterile Bulk 200 in. 20 bags of 10
21-381-093 10 mL Sterile Individually packed 100/Cs.
21-381-094 10 mL Non-sterile Bulk 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
This item is not returnable. View return policy
Catalog No. 21-385-124 Supplier VistaLab™ Technologies Supplier No. 30541008
This item is not returnable. View return policy

Unique trough-within-trough design maximizes reagent recovery

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Certifications/Compliance Pyrogen-free
Color White
Capacity (Metric) 1800 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Packaging Individually Wrapped
Quantity Each
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.