Learn More
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets
SDP
Catalog No. 21349B Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
121 to 200 μL
201 to 350 μL
31 to 120 μL
351 to 1000 μL
5 to 30 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-349B 31 to 120 μL
21-349A 5 to 30 μL
21-349C 121 to 200 μL
21-349D 201 to 350 μL
21-349E 351 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-349B Supplier VistaLab™ Technologies Supplier No. 9032
Add to Cart
Add to Cart
Provide materials for maintenance of MLA D-Tipper™, Precision, and two-stroke pipetters
Each kit contains seals and instructions
Order Info
Lubricant sold separately (P/N 22-146-318)
Specifications
For Use With Pipettes
Material Plastic
Volume (Metric) 31 to 120 μL
For Use With (Equipment) 31 to 120 μL MLA™ D-Tipper™ and Precision Pipette
Includes Lubricant, seals and instructions
Quantity 1 Ea.
Length (English) 2 in.
Width (English) 1 in.
Height (English) 1 in.