Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Visual Management Bouffant Cap Holder
SDP

Catalog No. 22000446
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-446 Keyhole Slots
22-000-445 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-446 Supplier Poltex Supplier No. BOUFFCAP2VMW
Add to Cart
Add to Cart

Visually manage and dispense PPE.

Durable and cleanable PETG construction

Specifications

Product Type PPE Dispenser
Color Clear
Width (Metric) 298 mm
Height (Metric) 520 mm
Material PETG
Mounting Keyhole Slots
For Use With (Application) Bouffant caps
Quantity 1 Ea.

Consumables not included.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.