Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ VisUBlock Mouse on Mouse Blocking Reagent
SDP

Catalog No. VB00101ML Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VB00101ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. VB00101ML Supplier R&D Systems™ Supplier No. VB00101ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Blocking reagent for use with primary antibodies from mouse host and mouse tissue.

  • Reduces background staining
  • Enables staining of mouse tissue with mouse IgG primary antibodies

Specifications

Content And Storage Store the unopened product at 2-8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 mL
Species Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.