Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Basic
SDP
Catalog No. VCTS021 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
VCTS021 50 mL
1 options
Catalog No. VCTS021 Supplier R&D Systems™ Supplier No. VCTS021
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
  • Unlike Universal solution, Acidic and Basic solutions may dramatically enhance immunoreactivity but may also affect tissue morphology.
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
Specifications
Content And Storage Store the unopened product at room temperature. Do not use past expiration date.
Product Line R&D Systems
Quantity 50 mL