Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Universal
SDP
Catalog No. VCTS023 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
VCTS023 50 mL
1 options
This item is not returnable. View return policy
Catalog No. VCTS023 Supplier R&D Systems™ Supplier No. VCTS023
Add to Cart
Add to Cart
This item is not returnable. View return policy
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
Specifications
Content And Storage Store the unopened product at room temperature. Do not use past expiration date.
Product Line R&D Systems
Quantity 50 mL