Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELITech Accu-Sed™ Plus ESR Controls with Peer Review
GSA_VASDP

Catalog No. 22024768 Shop All Clinical Data Products
Change view
Click to view available options
Type:
Abnormal Accu-Sed Plus ESR Control Kit
Accu-Sed Plus ESR Control Kit
Accu-Sed Plus ESR Control with Peer Review
Normal Accu-Sed Plus ESR Control Kit
Quantity:
2 x (1 x 8.5) mL
2 x (5 x 8.5) mL
5 x 8.5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-024-768 Accu-Sed Plus ESR Control Kit 2 x (5 x 8.5) mL
22-024-769 Normal Accu-Sed Plus ESR Control Kit 5 x 8.5 mL
22-024-770 Abnormal Accu-Sed Plus ESR Control Kit 5 x 8.5 mL
22-024-771 Accu-Sed Plus ESR Control with Peer Review 2 x (1 x 8.5) mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-024-768 Supplier ELITech Supplier No. DS71005A
Add to Cart
Add to Cart

VQC peer review program allows user to compare results to those of other laboratories running Clinical Data ESR systems

  • Designed specifically for Excyte™, Ves-Matic Automated ESR Testing Systems and Vital Diagnostics ESR Systems modified Westergren methods
  • Ready-to-uae bi-level control can be used directly from vial after mixing—no dilution or diluent required

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Compatible Methods Ves-Matic Automated ESR testing system and vital diagnostics ESR system modified Westergren methods
Content And Storage 15°C to 32°C
For Use With (Equipment) Excyte™ automated ESR testing system
Type Accu-Sed Plus ESR Control Kit
Intended Use Use in monitoring the performance of ESR procedures
Quantity 2 x (5 x 8.5) mL
Stability 31 Days Open, 18 Months Closed Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.