Learn More
Enzo Life Sciences Vitamin A acetate (1g). CAS: 127-47-9
SDP
Supplier:  Enzo Life Sciences ALX4600390001
Encompass_Preferred
Alternative name: Retinyl acetate. Purity: ≥95% (HPLC). Appearance: Off-white to pale yellow crystalline solid. Solubility: Soluble in ethanol, chloroform, and ether. Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.