Learn More
MONOBIND INC Vitamin B-12 AccuBind ELISA Kit - 96 wells
Supplier:  MONOBIND INC 7625300A
Encompass_Preferred
Vitamin B12 (cobalamin) is one of the 13 vitamins essential to human health and life. It is crucial for the construction of DNA, production of red blood cells and aids in the maintenance of a healthy nervous system. B12 deficiency damages the brain, spinal cord, peripheral nerves, and nerves of the eye. A B12 deficiency can result in symptoms ranging from severe anemia requiring blood transfusions, fatigue, muscle soreness, memory loss, and behavioral changes to serious and permanent nerve damage. B12 is found naturally in animal foods or is received through supplements, and must follow a complex pathway of several steps for proper absorption. Monobind's assay measures total B12 in serum.
Catalog No. NC1711222
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.