Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Vitamin B12a hydrochloride

Catalog No. AC462080050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462080050 5 g
AC462080010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462080050 Supplier Thermo Scientific Chemicals Supplier No. 462080050
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58288-50-9 | C62H91ClCoN13O15P | 1383.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58288-50-9
Molecular Formula C62H91ClCoN13O15P
Molecular Weight (g/mol) 1383.84
InChI Key BBQILPBAMXKFOB-UHFFFAOYNA-L
IUPAC Name λ²-cobalt(2+) 4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide hydrate hydrochloride
SMILES O.Cl.[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2[N-]C1=C(C)C1=NC(=CC3=NC(=C(C)C4=NC2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

Quantity 5 g
Formula Weight 1346.35
Chemical Name or Material Vitamin B12a hydrochloride
EINECSNumber 261-200-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.