Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin E, 1 mg = 1.1 IU, USP, 96-102%, Spectrum™ Chemical
SDP

Catalog No. 18607728 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-728 500 mL Amber Glass Bottle
18-607-725 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-728 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. VI135500ML

missing translation for 'validMainframeMsg'

VI135, 10191-41-0, C29H50O2

Spectrum™ Chemical Vitamin E, 1 mg = 1.1 IU, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

Acidity 1.0 mL
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 96 to 102%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Vanillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.