Learn More
STEMCELL Technologies Trolox, Size: 250 mg
SDP
Supplier:  STEMCELL Technologies 1000572
Encompass
Trolox is a cell-permeable and hydrophilic analog of vitamin E (Choe et al.; Lee et al.). In mice, Trolox has been shown to suppress osteoclast formation by downregulating receptor activator of NF-κB ligand (RANKL) induction and c-Fos expression (Lee et al.). Trolox also prevents cisplatin-induced apoptosis in renal epithelial cells (Xiao et al.).DIFFERENTIATION· Induces beta-like cell differentiation from human embryonic stem cells (Petersen et al.).CANCER RESEARCH· Enhances arsenic-mediated apoptosis in myeloma and breast cancer cells (Diaz et al.).
Catalog No. 50-206-9380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.