Learn More
Medchemexpress LLC HY-B2172 10mg Medchemexpress, Vitamin K CAS:12001-79-5 Purity:>98%
Supplier: Medchemexpress LLC HYB217210MG
Medchemexpress, HY-B2172 10mg Vitamin K CAS:12001-79-5 Vitamin K is a group of fat soluble vitamins, important for the function of numerous proteins within the body, such as the coagulation factors, osteocalcin and matrix-Gla protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.