Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin K1, MP Biomedicals™

Catalog No. MP21032832 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21032832 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21032832 Supplier MP Biomedicals, Inc Supplier No. 02103283.2
Add to Cart
Add to Cart

Used as a cofactor. Vitamin K1, MP Biomedicals a fat-soluble vitamin structurally similar to other forms of Vitamin K. It is a cofactor in the formation of coagulation factors II (prothrombin), VII, IX and X and of anticoagulant factors Protein C and S, a

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point -20°C (literature)
Density 0.984 (Lit.)
Boiling Point ∼140°C (literature)
Refractive Index 1.523-1.526 (at 25°C)
Quantity 250 mg
Solubility Information Soluble in chloroform (clear, yellow solution). Practically insoluble in water, sparingly soluble in alcohol, miscible with fatty oils.
Formula Weight 450.7
Physical Form Liquid
Recommended Storage Store at 4°C, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.