Learn More
Cayman Chemical DersImelagon 10mg
SDP
Supplier:  Cayman Chemical 4146810 MG
Encompass_Preferred
A fluorescent peptide ex/em 492/518 nm respectively has been used for site-specific labeling of target proteins via sortase-catalyzed transpeptidation
Catalog No. 50-297-8451
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.