Learn More
Ace Glass, Inc. VITON O RING SIZE 009 12 PK VITON O RING SIZE 009 12 PK
SDP
Supplier:  Ace Glass, Inc. 785507
Encompass_Preferred
Size 009 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. NC0192879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.