Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Viton O-ring for 47 mm In-Line Filter Holder, 2/pk

Catalog No. 502572482 Shop All Cytiva Products
Encompass

Viton O-ring for 47 mm In-Line Filter Holder, 2/pk

Catalog No. 50-257-2482 Supplier Cytiva Supplier No. 71242N
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Viton O-ring for 47 mm In-Line Filter Holder, 2/pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.