Learn More
Trajan Scientific and Medical™ Viton Sealing O-Ring for Shimadzu Liner
SDP
Catalog No. 0726533 Shop All SGE Inc. Products
Change view
Click to view available options
Quantity:
10/pack
Catalog No. Quantity
07-265-33 10/pack
1 options
Catalog No. 07-265-33 Supplier Trajan Scientific and Medical™ Supplier No. 0726533
Can be used at temperatures up to 300°C
Suitable for Shimadzu GCs
Specifications
For Use With (Application) Temperatures up to 300°C
For Use With (Equipment) SPL-2010 and SPL-2014 Injectors
Quantity 10/pack
Product Type Viton O-ring for Shimadzu GC's
Material Viton