Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vitronectin Monoclonal Antibody (A18)
GREENER_CHOICE

Catalog No. CSI0041802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CSI0041802 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CSI0041802 Supplier Invitrogen™ Supplier No. CSI0041802
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSI 004-18-02 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). CSI 004-18-02 cross-reacts with rabbit and horse vitronectin, no reactivity with human or sheep. CSI 004-18-02 has been successfully used in ELISA and Immunohistochemistry techniques. CSI 004-18-02 is suitable in ELISA and immunostaining of frozen PLP-fixed sections of bovine tissues. The antibody inhibits integrin-mediated cell adhesion to bovine vitronectin. It can be used to probe vitronectin conformation. CSI 004-18-02 reacts weakly in ELISA with vitronectin coated directly onto the microtiter plate. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitronectin
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A18
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene VTN
Gene Accession No. P22458
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Gene Symbols VTN
Host Species Mouse
Immunogen Lysed bovine corneal endothelial cells and extracellular matrix.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009128, 100059034, 507525
Target Species Rabbit, Bovine, Horse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.