Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Vitronectin (VTN-N) Recombinant Human Protein, Truncated

Catalog No. A31804
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL = 5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31804 10 mL = 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31804 Supplier Gibco™ Supplier No. A31804
Add to Cart
Add to Cart

Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs).

Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8 Medium (Cat. No. A1517001), VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.

  • Proven to maintain pluripotency and normal growth characteristics in multiple PSC lines when used with Essential 8 Medium
  • Supports PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages
  • Supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8 Medium
  • Optimized—designed in the laboratory of James Thomson for use with Essential 8 Medium
  • Defined—for reduced variability in your PSC cultures
  • Supports human pluripotent stem cell attachment and survival better than wild type vitronectin
  • Optimal environment for feeder-free pluripotent stem cell culture
  • Defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs)
TRUSTED_SUSTAINABILITY

Specifications

Concentration 500 μg/mL
For Use With (Application) Stem Cell Culture
Quantity 10 mL = 5 mg
Species Human
Recombinant Recombinant
Content And Storage Store at -80°C and protect from light.
Shipping Condition Dry Ice
Volume (Metric) 10 mL
Expression System E. coli
Protein Form Recombinant
Form Liquid
Protein Subtype Vitronectin
Research Category Stem Cell Research
Purity or Quality Grade Cell Culture Grade, Endotoxin-Screened
Show More Show Less
What substrate should I use with the PSC Definitive Endoderm Induction Kit?

We recommend using Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700), but Geltrex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301 or Cat. No. A1413302) can also be used.

Can pluripotent stem cells (PSCs) cryopreserved in a different culture condition be thawed and subsequently cultured in Gibco Essential 8 Medium and on vitronectin (VTN-N)?

Yes. PSCs cryopreserved from cultures of mTeSR Medium and BD Matrigel Basement Membrane Matrix may be thawed into Gibco Essential 8 Medium and plated on VTN-N. Certain lines may benefit from thawing into the medium and substrate they were growing in at the time of cryopreservation. Then at the next passage, use EDTA to passage the cells into Gibco Essential 8 Medium and VTN-N.

Can I use enzymes such as dispase and collagenase for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?

Enzymes such as dispase and collagenase do not work well with cells cultured in Gibco Essential 8 Medium on VTN-N. Use of these enzymes for passaging cells results in compromised viability and attachment.

What is the recommended passaging method to use with Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?

Cells cultured in Gibco Essential 8 Medium and VTN-N need to be passaged with EDTA.

What will my cells look like growing in Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?

You should expect to see normal pluripotent stem cell (PSC) morphology. The expected morphology of PSCs is demonstrated specifically by tightly packed colonies with defined borders and a high nucleus-to-cytoplasm ratio.

Can pluripotent stem cells (PSCs) previously cultured in other media and on other substrates be cultured in Gibco Essential 8 Medium and on Gibco Vitronectin?

Cells cultured in other feeder-free media systems, such as mTeSR Medium with Matrigel Basement Membrane Matrix, or StemPro hESC SFM with Geltrex Matrix, can be successfully cultured in Gibco Essential 8 Medium and VTN-N. In addition, PSCs grown on feeders with KnockOut SR have also been shown to be successfully cultured in Gibco Essential 8 Medium on VTN-N. However, when changing media systems, cells must be passaged either manually, or with EDTA prior to culturing in Gibco Essential 8 Medium on VTN-N.

Can Gibco Essential 8 Medium and Gibco Vitronectin (VTN-N) support long-term growth of pluripotent stem cells (PSCs)?

Gibco Essential 8 Medium and vitronectin have been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities, and maintain the ability of PSCs to differentiate into all three germ line lineages. As published by Chen et al (http://www.ncbi.nlm.nih.gov/pubmed/21478862) in the laboratory of James Thomson, the VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Gibco Essential 8 Medium.

Is Gibco Vitronectin (VTN-N) xeno-free (human or recombinant origin only)?

Yes. VTN-N is a defined, recombinant human protein.

What are the main differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems?

Here are three major differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems:

- Cells should be typically passaged ~24 hours sooner than they would be with other feeder-free media.
- Passaging should take place when cells are at ~85% confluency. If cells are passaged when they are more than 85% confluent, the health of the cells and final cell yield may be compromised.
- Cells must be passaged in EDTA. Collagenase and dispase are not recommended.

How does Gibco Vitronectin (VTN-N) compare to other basement membrane extracts?

Since VTN-N is a defined, recombinant human protein, variability is reduced in PSC cultures compared to human plasma-derived vitronectin and standard basement membrane extracts (BMEs). In addition, compared to full-length vitronectin and other defined substrates, VTN-N helps enable economical and scalable PSC culture.

What is Gibco Vitronectin (VTN-N)?

Gibco Vitronectin (VTN-N) is a recombinant, truncated human protein corresponding to the amino acid fragment 62-478 of human vitronectin expressed in E. coli. VTN-N is purified from inclusion bodies and refolded for use as a substrate for the feeder-free culture of human PSCs (http://www.ncbi.nlm.nih.gov/pubmed/21478862). When used with Gibco Essential 8 Medium, VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.

Can PSCs transitioned over into Essential 8 Medium on recombinant human laminin-521 (rhLaminin-521) from feeder-dependent cultures, then be transferred onto vitronectin (VTN-N)?

Yes. Following 2 passages on the rhLaminin-521 matrix, Versene or EDTA passaging should be used to subculture PSCs.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.