Learn More
Viva Products Vivacon 2-PCR Grade Centrifugal Concentrator (ETO treated)
Supplier: Viva Products VN02H43ETO
Centrifugal Ultrafiltration Device; Vivacon Model 2; For DNA concentration and desalting; Patented Hydrosart regenerated cellulose membrane for high recovery; Treated with dual-cycle ETO gas process to sterilize and denature DNA; Sample volume to 2 ml; Final concentrate volume as low as 55 microliters; Reverse spin for easy concentrate recovery; Fits fixed angle rotors for 15 ml (17 mm) conical bottom tubes; 125 mm length; 100,000 MWCO; 500/PK
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.