Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivapore 10

Catalog No. 01288105 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-288-105 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-288-105 Supplier Sartorius Supplier No. VPS010P00083
Add to Cart
Add to Cart

Vivapore 10 solvent absorption concentrators are disposable ultrafilters for macromolecule concentration from up to 10 mL aqueous samples.

  • Concentrate Equipment-Free: Solvent absorption is ideal where centrifuges are not available and for high-throughput applications.
  • Fill and Go: Simply add your sample and leave it to reach the desired final volume, with no risk of concentration to dryness.
  • Tailored Process Monitoring: Printed markings help to reduce the guesswork when each sample needs to reach its own specific concentration factor.
  • Maintained Sample Integrity: Self-contained, single use units ensure contamination-free concentration of individual samples.
  • Sensitive Molecule Friendly: Benchtop or refrigerated low pressure processing for shear and temperature sensitive molecules.
  • Expand or Exchange: Optional reservoirs provide the flexibility to double sample capacity or concentrate and desalt in one step
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Material (Membrane) PES
MWCO 7.5 kDa
Sample Volume 10 mL
Sterility Non-sterile
Type Solvent Absorption
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.