Learn More
Sartorius Vivapore Stand
Catalog No. 01288107 Shop All Sartorius Products
Change view
Click to view available options
Type:
Stand
Catalog No. Type
01-288-107 Stand
1 options
Catalog No. 01-288-107 Supplier Sartorius Supplier No. VPAST0001C
Add to Cart
Add to Cart
Vivapore stand can support up to four Vivapore 5 or 10 solvent absorption concentrators during use with simple push-fit handling.
  • Concentrate Equipment-Free: Solvent absorption is ideal where centrifuges are not available and for high-throughput applications.
  • Fill and Go: Simply add your sample and leave it to reach the desired final volume, with no risk of concentration to dryness.
  • Tailored Process Monitoring: Printed markings help to reduce the guesswork when each sample needs to reach its own specific concentration factor.
  • Maintained Sample Integrity: Self-contained, single use units ensure contamination-free concentration of individual samples.
  • Sensitive Molecule Friendly: Benchtop or refrigerated low pressure processing for shear and temperature sensitive molecules.
  • Expand or Exchange: Optional reservoirs provide the flexibility to double sample capacity or concentrate and desalt in one step
TRUSTED_SUSTAINABILITY
Specifications
Material ABS
Sterility Non-sterile
Type Stand