Learn More
Description
- Tame Tough Samples: Avoid membrane blocking with effective ultrafiltration for high protein or particle loaded samples.
- More to Analyze: Ultra-thin membranes ensure highly efficient analyte passage without adsorption or trapping.
- Reliable separation removes macromolecules that might otherwise interfere with downstream analysis.
- Label Friendly: A long outer surface with a consistent outer diameter allows barcoding in automated processes.
- With no cap and no disassembly required, the entire filtered sample can be retrieved directly with a pipette.
Specifications
Specifications
| Filter Type | Centrifuge Filter |
| Format | Spin Column |
| Material | PES |
| Material (Membrane) | PES |
| MWCO | 100 kDa |
| Sterility | Non-sterile |
| Type | Centrifugal |
| Volume (Metric) | 2.5 mL |
| Sample Volume | 2.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.