Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaspin™ Turbo 15 RC Ultrafiltration Centrifugal Concentrators

Catalog No. 14558877 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
100 kDa
30 kDa
Quantity:
12/Pk.
48/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14-558-877 100 kDa 12/Pk.
14-558-873 30 kDa 12/Pk.
14-558-874 30 kDa 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-558-877 Supplier Sartorius Supplier No. VS15TR41
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vivaspin™ Turbo 15 Regenerated Cellulose (RC) ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4-15 mL sample volumes. Acquire the fastest sample concentration with the highest recoveries for your protein samples.

Vivaspin™ Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs and market unique membrane choice of RC and PES membrane.

The Vivaspin™ Turbo 15 RC concentrators are the Regenerated Cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 μL.

  • Unique optimal membrane choice (PES and RC)
  • Complete recoveries
  • Fastest concentrations
  • Highest chemical resistance
  • Easy retentate collection
  • Graduated and markable
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Sterility Non-sterile
Housing Material Polypropylene
Quantity 12/Pk.
Type Centrifugal Concentrator
Volume (Metric) 15 mL
Length (Metric) 77 mm
For Use With (Equipment) Centrifuge
Sample Volume 15 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.