Learn More
Description
- Fastest sample concentration with maximum target molecule recoveries
- Simplest, pipette-friendly retentate retrieval
- Highest chemical and temperature resistance
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Spin Column |
| For Use With (Equipment) | Centrifuge |
| Material | SBC |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 10 kDa |
| Sample Volume | 4 to 15 mL |
| Sterility | Non-sterile |
| Quantity | 48 Pk. |
| Type | Centrifugal |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.