Learn More
Tocris Bioscience™ VKGILS-NH2
GSA_VAGREENER_CHOICE
Catalog No. 33925
Change view
Click to view available options
Quantity:
5 mg
Catalog No. Quantity
33-925 5 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-925 Supplier Tocris Bioscience™ Supplier No. 3392/5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Control peptide for SLIGKV-NH2 (Cat. No. 3010)
VKGILS-NH2 is a reversed amino acid sequence control peptide for SLIGKV-NH2, a protease-activated receptor 2 (PAR2) agonist.
Active Analog also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 5 mg