Learn More
Medchemexpress LLC Vlx600 | 327031-55-0 | MFCD01305422 | 317.35 g·mol⁻¹ | 5mg
Supplier: Medchemexpress LLC HY124065MG
VLX600 is an iron-chelating inhibitor of oxidative phosphorylation (OXPHOS) VLX600 causes mitochondrial dysfunction and induces a strong shift to glycolysis VLX600 displays selective cytotoxic activity against malignant cell and induces autophagy Anticancer activity[1][2]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.