Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VN1R5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116280
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116280 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116280 Supplier Invitrogen™ Supplier No. PA5116280
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VN1R5 Polyclonal Antibody for Western Blot

Putative pheromone receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VN1R5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene VN1R5
Gene Accession No. Q7Z5H4
Gene Alias G-protein coupled receptor GPCR26; hGPCR26; pheromone receptor; V1RL5; V1R-like 5; V1r-like receptor 5; VN1R5; vomeronasal 1 receptor 5 (gene/pseudogene); vomeronasal type-1 receptor 5
Gene Symbols VN1R5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 214-241 amino acids from the Central region of human VN1R5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317705
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.