Learn More
Med Vet International Medi-Vac Suction Tubing, 3/16 Inch Inner Diameter, 6 Foot Length, N56A, Each
Supplier:  Med Vet International 251161EA
Encompass_Preferred
Resists collapse from pressure, up to 22 Hg vacuum Single use, sterile Maxi-Grip™ connectors Rigid male connector
Catalog No. 50-304-4923
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.