Learn More
Invitrogen™ VNN2 Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™
Description
Specificity: Human VNN2/Vanin-2
Specifications
Specifications
| Antigen | VNN2 |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 018 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | VNN2 |
| Gene Accession No. | O95498 |
| Gene Alias | FOAP-4; Glycosylphosphatidyl inositol-anchored protein GPI-80; GPI-80; LOW QUALITY PROTEIN: vascular non-inflammatory molecule 2; pantetheinase; Protein FOAP-4; vanin 2; Vanin-2; Vannin 2; vascular non-inflammatory molecule 2; VNN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.