Learn More
Medchemexpress LLC Vnt-101 | 2894060-67-2 | 456.41 | C21H23F3N2O6 | 10 MG

Supplier: Medchemexpress LLC HY17546910MG
VNT-101 is a small-molecule, orally active inhibitor of influenza A virus nucleoprotein (NP) oligomerization. It disrupts NP-NP protein-protein interactions to destabilize the viral ribonucleoprotein complex and has been used in vitro and in vivo to study antiviral activity across multiple influenza A subtypes.
- Disrupts nucleoprotein oligomerization to inhibit viral replication.
- Orally bioavailable small molecule suitable for in vivo studies.
- Demonstrated activity against multiple influenza A subtypes.
- Molecular weight 456.41 and molecular formula C21H23F3N2O6.
- CAS number 2894060-67-2 for substance identification.
- Supplied as a solid for research use with supporting characterization data.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.