Learn More
Apexbio Technology LLC VO-Ohpic trihydrate 476310-60-8 25mg
Supplier: Apexbio Technology LLC A392325
VO-Ohpic trihydrate (CAS 476310-60-8) is a highly selective small-molecule inhibitor of the lipid phosphatase PTEN (phosphatase and tensin homolog) with an in vitro IC50 of 35 nM VO-Ohpic shows marked selectivity for PTEN over other phosphatases including CBPs SopB myotubularin SAC1 and PTP- In cell-based assays VO-Ohpic induces dose-dependent phosphorylation of Akt at Ser473 and Thr308 saturating at 75 nM and promotes Akt functional activation as indicated by decreased FoxO3a transcriptional activity In vivo VO-Ohpic administration in MDA PCa-2b tumor xenograft mice results in reduced tumor growth and increased survival This compound serves as a valuable tool for investigating PTEN function and its modulation in insulin signaling glucose uptake and cancer research
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.