Learn More
Apexbio Technology LLC Vortioxetine 508233-74-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A39265.1
Encompass_Preferred
Vortioxetine (CAS 508233-74-7) is a multimodal antidepressant currently under investigation for the treatment of depression It demonstrates high affinity for recombinant human serotonin receptors including 5-HT1A (Ki 15 nM) 5-HT1B (Ki 33 nM) 5-HT3A (Ki 3 7 nM) 5-HT7 (Ki 19 nM) noradrenergic 1 receptors (Ki 46 nM) and the serotonin transporter SERT (Ki 1 6 nM) Vortioxetine acts as an antagonist at 5-HT3A and 5-HT7 partial agonist at 5-HT1B agonist at 5-HT1A and exhibits potent SERT inhibition In vivo studies show that short-term administration increases extracellular 5-HT in rat brain Vortioxetine s multimodal activity makes it a valuable tool for research into serotonergic modulation and antidepressant mechanisms
Catalog No. 50-000-96265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.