Learn More
Selleck Chemical LLC Volasertib S2235-100mg
SDP
Supplier:  Selleck Chemical LLC S2235100MG
Encompass_Preferred
Volasertib is a highly potent Plk1 inhibitor with IC50 of 0 87 nM in a cell-free assay It shows 6- and 65-fold greater selectivity against Plk2 and Plk3 Volasertib induces cell cycle arrest and apoptosis in various cancer cells Phase 3
Catalog No. 50-311-2788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.